Learn More
Grainger PHYSICIANSCARE Single-Use Eyewash Bottle
Catalog No. NC2244550
Encompass_Preferred
The PhysiciansCare Eyewash Bottle offers a portable, sterile saline solution for emergency eye rinsing. With a twist-off cap and soft nozzle, it's ideal for worksites and schools, providing 15 minutes of irrigation without plumbing. ANSI compliant with a 5-year shelf life, it's perfect for immediate first aid until professional care is available.
Catalog No. NC2244550 Supplier Grainger Supplier No. 36N058
The PhysiciansCare Eyewash Bottle offers a portable, sterile saline solution for emergency eye rinsing. With a twist-off cap and soft nozzle, it's ideal for worksites and schools, providing 15 minutes of irrigation without plumbing. ANSI compliant with a 5-year shelf life, it's perfect for immediate first aid until professional care is available.