Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Unibor™ Single-Use Gel Hand Sanitizer Packets

Catalog No. 19350100
Change view
Click to view available options
Quantity:
100 Packets
30 Packets
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
19-350-100 30 Packets
19-350-101 100 Packets
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
  • Not for consumption
  • Flammable
Catalog No. 19-350-100 Supplier Unibor™ Supplier No. UHS030
Add to Cart
Add to Cart
This item is not returnable. View return policy
  • Not for consumption
  • Flammable

Innovative one-time use packets of high quality gel sanitizer. More effective than wipes, with substantially less environmental impact.

  • Great smelling hand sanitizer that is safe, effective, and soft on skin
  • The perfect amount of sanitizer in each packet
  • For personal use in wallets, purses, and pockets
  • For business/organizational use as complimentary germ-prevention for customers and guests
  • Ideal for schools and school cafeterias, provide one packet with every lunch
  • Environmentally sound - removes need for plastic bottles, and wipes that take hundreds of years to biodegrade

Specifications

Product Type Hand Sanitizer Gel
Color Clear
Certifications/Compliance FDA Registered OTC Monograph, Part333A
For Use With (Application) Eliminating Germs on Hands
Packaging Single-Use Packet
Quantity 30 Packets
Volume (Metric) 1.5 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.