Learn More
Unibor™ Single-Use Gel Hand Sanitizer Packets
Description
- Great smelling hand sanitizer that is safe, effective, and soft on skin
- The perfect amount of sanitizer in each packet
- For personal use in wallets, purses, and pockets
- For business/organizational use as complimentary germ-prevention for customers and guests
- Ideal for schools and school cafeterias, provide one packet with every lunch
- Environmentally sound - removes need for plastic bottles, and wipes that take hundreds of years to biodegrade
Specifications
Specifications
| Product Type | Hand Sanitizer |
| Color | Clear |
| Form | Gel |
| Certifications/Compliance | FDA Registered OTC Monograph, Part333A |
| For Use With (Application) | Eliminating Germs on Hands |
| Packaging | Single-Use Packet |
| Quantity | 100 Packets |
| Volume (Metric) | 1.5 mL |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.