Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CoaguSense™ Single Use Lancets

Catalog No. 23654004
Change view
Click to view available options
Quantity:
100
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
23-654-004 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 23-654-004 Supplier CoaguSense™ Supplier No. 03P5804
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

CoaguSense™ Single Use Lancets are 21 gauge and designed for good PT/INR blood flow.

  • Box of 100
  • 21 gauge lancets

Specifications

Content And Storage Room Temperature
Format Lancets
Type Single Use
Quantity 100
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.