Learn More
BURKLE INC Single-use liquid funnel Bio-PE, Ø 3.94 in. (100 mm), white
Catalog No. 503081213
Encompass_Preferred
Disposable funnel for reliable filling of liquids, ideal for filling bottles and other containers with a narrow filler opening.
The disposable funnel ensures no loss when transferring liquid samples or free-flowing solids. That makes this practical and hygienic pouring aid a helpful accessory for sampling or in laboratories.
Clean room produced funnel with suspension eye and venting fins for ventilation to ensure uniform flow while pouring.
In combination with a filter paper the disposable funnel is also well suited for filtering.
Catalog No. 50-308-1213 Supplier BURKLE INC Supplier No. 53790040
Disposable funnel for reliable filling of liquids, ideal for filling bottles and other containers with a narrow filler opening.
The disposable funnel ensures no loss when transferring liquid samples or free-flowing solids. That makes this practical and hygienic pouring aid a helpful accessory for sampling or in laboratories.
Clean room produced funnel with suspension eye and venting fins for ventilation to ensure uniform flow while pouring.
In combination with a filter paper the disposable funnel is also well suited for filtering.