Learn More
Electron Microscopy Sciences Single-Use Notched Forceps, 10 mm with 0.25 mm Notch Tips, 80 mm Handle
SDP
Catalog No. 501931095
Encompass_Preferred
These unique sterile forceps have a plastic base and tips made of are 316 stainless steel.
Tips: 10 mm with 0.25 mm Notch
Handle: 80 mm
OAL: 100 mm
Catalog No. 50-193-1095 Supplier Electron Microscopy Sciences Supplier No. 7793793
These unique sterile forceps have a plastic base and tips made of are 316 stainless steel.
Tips: 10 mm with 0.25 mm Notch
Handle: 80 mm
OAL: 100 mm