Learn More
Honeywell™ Single-Use Powder Free Nitrile Gloves
Catalog No. 19085581
Change view
Click to view available options
Size:
Small
X-Large
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-085-581 X-Large
19-085-578 Small
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 19-085-581 Supplier Honeywell™ Supplier No. 4580386XL
This item is not returnable. View return policy
High-quality nitrile gloves that are certified for food handling.
  • AQL 1.5 and food handling certified
  • Guarantees a high level of quality and contact with foodstuffs (non-damaging to health, glove components do not migrate toward food)
  • Nitrile glove apply to those allergic to latex
  • Very fine touch
TRUSTED_SUSTAINABILITY
Specifications
Color Blue
Disposable Single-use
Material Nitrile
Size X-Large
Thickness (English) 4 mil
Thickness (Metric) 0.10 mm
Length (English) 9.5 in.