Learn More
BURKLE INC Single-use powder funnel Bio-PE, Ø 3.94 in. (100 mm), white
Catalog No. 503081377
Encompass_Preferred
The disposable powder funnel has a short, wide discharge and is suitable for transferring powdered samples and materials, granulates and suspensions as well as other liquids.
The wide opening of the disposable powder funnel prevents the powder or granulates from clogging while dispensing. That makes this practical and hygienic pouring aid a helpful accessory for sampling or in laboratories.
In combination with a filter paper the disposable funnel is also well suited for filtering.
Catalog No. 50-308-1377 Supplier BURKLE INC Supplier No. 53790041
The disposable powder funnel has a short, wide discharge and is suitable for transferring powdered samples and materials, granulates and suspensions as well as other liquids.
The wide opening of the disposable powder funnel prevents the powder or granulates from clogging while dispensing. That makes this practical and hygienic pouring aid a helpful accessory for sampling or in laboratories.
In combination with a filter paper the disposable funnel is also well suited for filtering.