Learn More
Pendotech Pendotech 1 Inch Hose Barb Single Use Pressure Sensor
Catalog No. NC1618832
Encompass
This item has a minimum order quantity of 5 per supplier requirements
Single Use Pressure Sensor, non-sterile, polysulfone 1 inch hose barb
Catalog No. NC1618832 Supplier Pendotech Supplier No. PREPSN100
This item has a minimum order quantity of 5 per supplier requirements
Single Use Pressure Sensor, non-sterile, polysulfone 1 inch hose barb