Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ CytoQuest™ CR: Plugs Set

Catalog No. 89139311 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 Set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-139-311 1 Set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-139-311 Supplier Abnova™ Supplier No. KA4441
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Includes

Each set contains 20 pieces each of: Inlet plug (blue), outlet plug (white) and outlet plug (blue) with tubing.

Single use, replaceable plugs to avoid sample cross contamination on CytoQuest CR

  • For use with CytoQuest CR
Applications:
Immunofluorescence

Recommended Storage

Store at room temperature.

Specifications

Content And Storage Store at room temperature.
Includes Each set contains 20 pieces each of: Inlet plug (blue), outlet plug (white) and outlet plug (blue) with tubing
Quantity 1 Set
For Use With (Equipment) CytoQuest CR

For research use only (RUO).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.