Learn More
BURKLE INC DispoPipette LaboPlast, HDPE, 19.69 in. (500 mm), 3.38 oz. (100 ml)
Catalog No. 503081324
Encompass_Preferred
Simple disposable sampler for taking samples of low viscosity media. If the DispoPipette is inserted into the liquid opened, an all-layer sample is obtained. If it is closed while it is inserted up to the sampling point and then opened, a target-point sample is obtained. Very easy to operate. With two opening sizes (2.5/5.5 mm (0.10/0.22 in.)) for different viscosities.
Catalog No. 50-308-1324 Supplier BURKLE INC Supplier No. 53935500
Simple disposable sampler for taking samples of low viscosity media. If the DispoPipette is inserted into the liquid opened, an all-layer sample is obtained. If it is closed while it is inserted up to the sampling point and then opened, a target-point sample is obtained. Very easy to operate. With two opening sizes (2.5/5.5 mm (0.10/0.22 in.)) for different viscosities.