Learn More
BURKLE INC DispoLadle LaboPlast, HDPE, 19.69 in. (500 mm)
Catalog No. 503081287
Encompass_Preferred
DispoLadle is a disposable sampling scoop specially designed for accurate liquid sampling. The scaled scoop features embossed measurements in 10 ml (0.34 oz.) increments, up to 200 ml (6.76 oz.), ensuring precise volume control.
Cleanroom manufactured and individually packaged, DispoLadle meets strict hygiene standards. Its ergonomic handle and dual spouts allow easy handling for both right- and left-handed users, making liquid sampling simple and efficient.
Catalog No. 50-308-1287 Supplier BURKLE INC Supplier No. 53936640
DispoLadle is a disposable sampling scoop specially designed for accurate liquid sampling. The scaled scoop features embossed measurements in 10 ml (0.34 oz.) increments, up to 200 ml (6.76 oz.), ensuring precise volume control.
Cleanroom manufactured and individually packaged, DispoLadle meets strict hygiene standards. Its ergonomic handle and dual spouts allow easy handling for both right- and left-handed users, making liquid sampling simple and efficient.