Learn More
BURKLE INC DispoTube LaboPlast, HDPE, 19.69 in. (500 mm), 3.38 oz. (100 ml)
Catalog No. 503081331
Encompass_Preferred
The ergonomic handle of the DispoTube makes it easier to work and facilitates a good grip even under difficult conditions, for example with gloves. Disposable sampler for taking samples of liquids.
Catalog No. 50-308-1331 Supplier BURKLE INC Supplier No. 53936620
The ergonomic handle of the DispoTube makes it easier to work and facilitates a good grip even under difficult conditions, for example with gloves. Disposable sampler for taking samples of liquids.