Learn More
BURKLE INC DispoTube SteriPlast, HDPE, sterile, 39.37 in. (1000 mm), 6.76 oz. (200 ml)
Catalog No. 503081335
Encompass_Preferred
The ergonomic handle of the DispoTube makes it easier to work and facilitates a good grip even under difficult conditions, for example with gloves. Disposable sampler for taking samples of liquids.
Catalog No. 50-308-1335 Supplier BURKLE INC Supplier No. 53936631
The ergonomic handle of the DispoTube makes it easier to work and facilitates a good grip even under difficult conditions, for example with gloves. Disposable sampler for taking samples of liquids.