Learn More
BURKLE INC LiquiDispo SteriPlast, HDPE, sterile, 19.69 in. (500 mm), 3.38 oz. (100 ml)
Catalog No. 503081333
Encompass_Preferred
The LiquiDispo disposable sampler is cleanroom-manufactured, gamma-ray sterilized and individually packaged to ensure contamination-free sampling in sensitive environments. The ergonomic handle provides comfortable, precise operation, making it easy to collect liquid samples quickly and safely. Ideal for use in the food industry, pharmaceutical production, and laboratory applications where hygiene, accuracy, and safety are critical.
Catalog No. 50-308-1333 Supplier BURKLE INC Supplier No. 53931121
Add to Cart
Add to Cart
The LiquiDispo disposable sampler is cleanroom-manufactured, gamma-ray sterilized and individually packaged to ensure contamination-free sampling in sensitive environments. The ergonomic handle provides comfortable, precise operation, making it easy to collect liquid samples quickly and safely. Ideal for use in the food industry, pharmaceutical production, and laboratory applications where hygiene, accuracy, and safety are critical.