Learn More
BURKLE INC LiquiDispo LaboPlast, HDPE, 19.69 in. (500 mm), 3.38 oz. (100 ml)
Catalog No. 503081349
Encompass_Preferred
The LiquiDispo disposable sampler is cleanroom-manufactured and individually packaged to ensure contamination-free sampling in sensitive environments. The ergonomic handle provides comfortable, precise operation, making it easy to collect liquid samples quickly and safely. Ideal for use in the food industry, pharmaceutical production, and laboratory applications where hygiene, accuracy, and safety are critical.
Catalog No. 50-308-1349 Supplier BURKLE INC Supplier No. 53931120
The LiquiDispo disposable sampler is cleanroom-manufactured and individually packaged to ensure contamination-free sampling in sensitive environments. The ergonomic handle provides comfortable, precise operation, making it easy to collect liquid samples quickly and safely. Ideal for use in the food industry, pharmaceutical production, and laboratory applications where hygiene, accuracy, and safety are critical.