Learn More
BURKLE INC PowderDispo SteriPlast, HDPE, sterile, 11.81 in. (300 mm), 2.03 ml (60 ml)
Catalog No. 503081300
Encompass_Preferred
PowderDispo is the ideal disposable sampler for cross-section samples of powdery or slightly cohesive substances. A complete all-layer sample can be taken from bags or smaller containers with the lance.
Catalog No. 50-308-1300 Supplier BURKLE INC Supplier No. 53935511
PowderDispo is the ideal disposable sampler for cross-section samples of powdery or slightly cohesive substances. A complete all-layer sample can be taken from bags or smaller containers with the lance.