Learn More
BURKLE INC PowderDispo LaboPlast, HDPE, 11.81 in. (300 mm), 2.03 oz. (60 ml)
Catalog No. 503081327
Encompass_Preferred
PowderDispo is the ideal disposable sampler for cross-section samples of powdery or slightly cohesive substances. A complete all-layer sample can be taken from bags or smaller containers with the lance.
Catalog No. 50-308-1327 Supplier BURKLE INC Supplier No. 53935510
PowderDispo is the ideal disposable sampler for cross-section samples of powdery or slightly cohesive substances. A complete all-layer sample can be taken from bags or smaller containers with the lance.