Learn More
BURKLE INC PowderDispo LaboPlast, HDPE, 19.69 in. (500 mm), 3.38 oz. (100 ml)
Catalog No. 503081343
Encompass_Preferred
PowderDispo is the ideal disposable sampler for cross-section samples of powdery or slightly cohesive substances. A complete all-layer sample can be taken from bags or smaller containers with the lance.
Catalog No. 50-308-1343 Supplier BURKLE INC Supplier No. 53935522
PowderDispo is the ideal disposable sampler for cross-section samples of powdery or slightly cohesive substances. A complete all-layer sample can be taken from bags or smaller containers with the lance.