Learn More
BURKLE INC MicroDispo LaboPlast, HDPE, 39.37 in. (1000 mm), 0.34 oz. (10 ml)
Catalog No. 503081282
Encompass_Preferred
Quite small sample quantities (10 ml (0.34 oz.)) can be taken with MicroDispo. Especially in pharmaceutical applications it often happens that only a small quantity can or should be extracted for analysis. Using the disposable sampler, precise target point samples can be taken of free-flowing powders and small-grain bulk materials.
Catalog No. 50-308-1282 Supplier BURKLE INC Supplier No. 53932250
Quite small sample quantities (10 ml (0.34 oz.)) can be taken with MicroDispo. Especially in pharmaceutical applications it often happens that only a small quantity can or should be extracted for analysis. Using the disposable sampler, precise target point samples can be taken of free-flowing powders and small-grain bulk materials.