Learn More
BURKLE INC DispoLance SteriPlast, HDPE, sterile, 19.69 in. (500 mm)
Catalog No. 503081345
Encompass_Preferred
The DispoLance disposable sampler can be used for taking large and small sample quantities quickly and easily, directly from bags. The sampler is suitable for free-flowing powders and small-grain granulates.
The DispoLance sampler pierces into the container with the sturdy tip and the sample flows through the open inner tube directly into the container or bag that is held underneath.
Catalog No. 50-308-1345 Supplier BURKLE INC Supplier No. 53935571
The DispoLance disposable sampler can be used for taking large and small sample quantities quickly and easily, directly from bags. The sampler is suitable for free-flowing powders and small-grain granulates.
The DispoLance sampler pierces into the container with the sturdy tip and the sample flows through the open inner tube directly into the container or bag that is held underneath.