Learn More
BURKLE INC TargetDispo LaboPlast, HDPE, 19.69 in. (500 mm), 0.57 oz. (17 ml)
Catalog No. 503081292
Encompass_Preferred
TargetDispo is used to take a target point sample at a position in the sample material that has previously been precisely determined. The sampler pierces into the powder or granulate while closed. When it reaches the sample removal point it is opened and then closed again. In this way a precise target-point sample is obtained. The sample can simply be emptied through the open handle.
Catalog No. 50-308-1292 Supplier BURKLE INC Supplier No. 53934460
TargetDispo is used to take a target point sample at a position in the sample material that has previously been precisely determined. The sampler pierces into the powder or granulate while closed. When it reaches the sample removal point it is opened and then closed again. In this way a precise target-point sample is obtained. The sample can simply be emptied through the open handle.