Learn More
BURKLE INC TargetDispo SteriPlast, HDPE steril, 39.37 in. (1000 mm), 0.57 oz. (17 ml)
Catalog No. 503081339
Encompass_Preferred
TargetDispo is used to take a target point sample at a position in the sample material that has previously been precisely determined. The sampler pierces into the powder or granulate while closed. When it reaches the sample removal point it is opened and then closed again. In this way a precise target-point sample is obtained. The sample can simply be emptied through the open handle.
Catalog No. 50-308-1339 Supplier BURKLE INC Supplier No. 53934471
TargetDispo is used to take a target point sample at a position in the sample material that has previously been precisely determined. The sampler pierces into the powder or granulate while closed. When it reaches the sample removal point it is opened and then closed again. In this way a precise target-point sample is obtained. The sample can simply be emptied through the open handle.