Learn More
BURKLE INC DispoPicker SteriPlast, HDPE sterile, 19.69 in. (500 mm), 3.38 oz. (100 ml)
Catalog No. 503081301
Encompass_Preferred
Disposable sampler for powders and small-grain bulk materials. The sterile, cleanroom-manufactured DispoPicker ensures contamination-free sampling in sensitive environments. Because the tip is closed, the sample material cannot trickle back out of the sample lance, preserving sample integrity. Ideal for cross-section samples in pharmaceutical, food, and chemical applications.
Catalog No. 50-308-1301 Supplier BURKLE INC Supplier No. 53935551
Disposable sampler for powders and small-grain bulk materials. The sterile, cleanroom-manufactured DispoPicker ensures contamination-free sampling in sensitive environments. Because the tip is closed, the sample material cannot trickle back out of the sample lance, preserving sample integrity. Ideal for cross-section samples in pharmaceutical, food, and chemical applications.