Learn More
BURKLE INC DispoPicker SteriPlast, HDPE, sterile, 11.81 in. (300 mm), 2.03 ml (60 ml)
Catalog No. 503081329
Encompass_Preferred
Disposable sampler for powders and small-grain bulk materials. The sterile, cleanroom-manufactured DispoPicker ensures contamination-free sampling in sensitive environments. Because the tip is closed, the sample material cannot trickle back out of the sample lance, preserving sample integrity. Ideal for cross-section samples in pharmaceutical, food, and chemical applications.
Catalog No. 50-308-1329 Supplier BURKLE INC Supplier No. 53935541
Disposable sampler for powders and small-grain bulk materials. The sterile, cleanroom-manufactured DispoPicker ensures contamination-free sampling in sensitive environments. Because the tip is closed, the sample material cannot trickle back out of the sample lance, preserving sample integrity. Ideal for cross-section samples in pharmaceutical, food, and chemical applications.