Learn More
BURKLE INC DispoLance LaboPlast, HDPE, 11.81 in. (300 mm)
Catalog No. 503081346
Encompass_Preferred
The DispoLance disposable sampler can be used for taking large and small sample quantities quickly and easily, directly from bags. The sampler is suitable for free-flowing powders and small-grain granulates.
The DispoLance sampler pierces into the container with the sturdy tip and the sample flows through the open inner tube directly into the container or bag that is held underneath.
Catalog No. 50-308-1346 Supplier BURKLE INC Supplier No. 53935560
The DispoLance disposable sampler can be used for taking large and small sample quantities quickly and easily, directly from bags. The sampler is suitable for free-flowing powders and small-grain granulates.
The DispoLance sampler pierces into the container with the sturdy tip and the sample flows through the open inner tube directly into the container or bag that is held underneath.