Learn More
BURKLE INC ZoneDispo LaboPlast, HDPE, 39.37 in. (1000 mm), 6.76 oz. (200 ml)
Catalog No. 503081228
Encompass_Preferred
The ZoneDispo disposable all-layer sampler is used in sampling to collect a representative all-layer sample from three different layers. Suitable for free-flowing powders and small-grain granulates. The collected sample can be secured and transported in the closed sampler.
Catalog No. 50-308-1228 Supplier BURKLE INC Supplier No. 53934430
The ZoneDispo disposable all-layer sampler is used in sampling to collect a representative all-layer sample from three different layers. Suitable for free-flowing powders and small-grain granulates. The collected sample can be secured and transported in the closed sampler.