Learn More
BURKLE INC MultiDispo SteriPlast, HDPE sterile, 39.37 in. (1000 mm), 6.76 oz. (200 ml)
Catalog No. 503081291
Encompass_Preferred
Disposable zone samplers designed for sampling multiple layers of free-flowing powders and small-grain granulates. Manufactured in a cleanroom environment, individually packaged, and gamma-sterilized to meet the highest standards of hygiene and contamination-free use.
The representative all-layer sample can be easily emptied directly through the open handle into your sample container, making it ideal for precise and efficient material analysis in pharmaceutical, food, and chemical industries.
Catalog No. 50-308-1291 Supplier BURKLE INC Supplier No. 53934451
Disposable zone samplers designed for sampling multiple layers of free-flowing powders and small-grain granulates. Manufactured in a cleanroom environment, individually packaged, and gamma-sterilized to meet the highest standards of hygiene and contamination-free use.
The representative all-layer sample can be easily emptied directly through the open handle into your sample container, making it ideal for precise and efficient material analysis in pharmaceutical, food, and chemical industries.