Learn More
BURKLE INC MultiDispo LaboPlast, HDPE, 19.69 in. (500 mm), 3.38 oz. (100 ml)
Catalog No. 503081338
Encompass_Preferred
Disposable zone samplers designed for sampling multiple layers of free-flowing powders and small-grain granulates. Manufactured in a cleanroom environment and individually packaged to ensure maximum hygiene and contamination-free use.
The representative all-layer sample can be easily emptied directly through the open handle into your sample container, making it ideal for precise and efficient material analysis in pharmaceutical, food, and chemical industries.
Catalog No. 50-308-1338 Supplier BURKLE INC Supplier No. 53934440
Disposable zone samplers designed for sampling multiple layers of free-flowing powders and small-grain granulates. Manufactured in a cleanroom environment and individually packaged to ensure maximum hygiene and contamination-free use.
The representative all-layer sample can be easily emptied directly through the open handle into your sample container, making it ideal for precise and efficient material analysis in pharmaceutical, food, and chemical industries.