Learn More
BURKLE INC ViscoDispo LaboPlast, HDPE, 39.37 in. (1000 mm), 6.76 oz. (200 ml)
Catalog No. 503081348
Encompass_Preferred
The ViscoDispo disposable sampler can be used to take samples of high-viscosity media up to 100,000 mPas. The cleanroom-manufactured and individually packaged design ensures safe and contamination-free sampling. The ergonomically shaped handle and the very stable drawbar make it easy to draw even viscous substances.
Catalog No. 50-308-1348 Supplier BURKLE INC Supplier No. 53933330
The ViscoDispo disposable sampler can be used to take samples of high-viscosity media up to 100,000 mPas. The cleanroom-manufactured and individually packaged design ensures safe and contamination-free sampling. The ergonomically shaped handle and the very stable drawbar make it easy to draw even viscous substances.