Learn More
BURKLE INC ViscoDispo SteriPlast, HDPE, sterile, 19.69 in. (500 mm), 3.38 oz. (100 ml)
Catalog No. 503081303
Encompass_Preferred
The ViscoDispo disposable sampler can be used to take samples of high-viscosity media up to 100,000 mPas. The cleanroom-manufactured, individually packaged, gamma-ray sterilized design ensures safe and contamination-free sampling. The ergonomically shaped handle and the very stable drawbar make it easy to draw even viscous substances.
Catalog No. 50-308-1303 Supplier BURKLE INC Supplier No. 53933321
The ViscoDispo disposable sampler can be used to take samples of high-viscosity media up to 100,000 mPas. The cleanroom-manufactured, individually packaged, gamma-ray sterilized design ensures safe and contamination-free sampling. The ergonomically shaped handle and the very stable drawbar make it easy to draw even viscous substances.