Learn More
Fine Science Tools Single Use Feeding Needles, 5/pk
SDP
Catalog No. NC9711926
Encompass
Product: Single Use Feeding Needles - Straight/38mm/20G/1.75mm Tip Diameter/ Gauge: 20/ Tip Diameter: 1.75mm/ Shape: Straight/ Tip Shape: Barrel/ Material: Composite/ Length: 38mm/ Sterile: Yes/ Style: Single Use/ Unit. These economical feeding needles feature plastic luer hubs with silicone tips. They are nontoxic and nonpyrogenic. Needles can be easily formed to any desired shape.
Catalog No. NC9711926 Supplier Fine Science Tools Supplier No. 1806120
Product: Single Use Feeding Needles - Straight/38mm/20G/1.75mm Tip Diameter/ Gauge: 20/ Tip Diameter: 1.75mm/ Shape: Straight/ Tip Shape: Barrel/ Material: Composite/ Length: 38mm/ Sterile: Yes/ Style: Single Use/ Unit. These economical feeding needles feature plastic luer hubs with silicone tips. They are nontoxic and nonpyrogenic. Needles can be easily formed to any desired shape.