Learn More
Pendotech Pendotech 1/4 Inch Hose Barb Single Use Uv Flow Cell
Catalog No. NC1380266
Encompass
Single use UV flow cell, 0.5cm path length, non-sterile, polysulfone, 1/4" inch hose barb
Catalog No. NC1380266 Supplier Pendotech Supplier No. SPECPSN025
Single use UV flow cell, 0.5cm path length, non-sterile, polysulfone, 1/4" inch hose barb