Learn More
Perkin Elmer US LLC Single Valve SPE Cartridge
Catalog No. 502111976
Encompass_Preferred
The online SPE cartridge has been specially designed for elution to the HPLC column. Dimensions are optimized to combine high extraction capacity with small elution volumes. Standard dimensions are 10 mm in length and 2 mm inner diameter, but 10 x 1 mm is also available.
Catalog No. 50-211-1976 Supplier Perkin Elmer US LLC Supplier No. N2991112
The online SPE cartridge has been specially designed for elution to the HPLC column. Dimensions are optimized to combine high extraction capacity with small elution volumes. Standard dimensions are 10 mm in length and 2 mm inner diameter, but 10 x 1 mm is also available.