Learn More
Electron Microscopy Sciences Conductive Black Single Wafer Container, 3", Polypropylene Material, 84 x 11 mm
SDP
Catalog No. 501929908
Encompass_Preferred
The carriers are used for storage and or shipping wafers. They prevent all types of damage and contamination to the wafer during shipping and even storage. Each of the containers are packed in 1000 class clean room and bag. The set includes the Container , the cover and the spring.
Use: 3" (75mm)
Material: Polypropylene
Dimensions: 84 x 11 mm
Catalog No. 50-192-9908 Supplier Electron Microscopy Sciences Supplier No. 649193
The carriers are used for storage and or shipping wafers. They prevent all types of damage and contamination to the wafer during shipping and even storage. Each of the containers are packed in 1000 class clean room and bag. The set includes the Container , the cover and the spring.
Use: 3" (75mm)
Material: Polypropylene
Dimensions: 84 x 11 mm