Learn More
Electron Microscopy Sciences Single Wash Bottle Holder
SDP
Catalog No. 5029690
Encompass_Preferred
These wash bottle holders protect bench top from spills and stains and provide access to solvent wash bottles. They are made from chemical resistant plastic that is easy to clean.
Catalog No. 50-296-90 Supplier Electron Microscopy Sciences Supplier No. 6437301
These wash bottle holders protect bench top from spills and stains and provide access to solvent wash bottles. They are made from chemical resistant plastic that is easy to clean.