Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Axygen™ Single Well High Profile Reagent Reservoirs
GREENER_CHOICE

Catalog No. 14222422 Shop All Axygen Products
Change view
Click to view available options
Capacity (Metric):
240 mL
282 mL
290 mL
Sterility:
Non-sterile
Sterile
Channels:
8
12
96
384
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric) Sterility Channels
14-222-422 290 mL Sterile 8
14-222-418 282 mL Sterile 384
14-222-412 290 mL Sterile 12
14-222-421 290 mL Non-sterile 8
14-222-411 290 mL Non-sterile 12
14-222-423 240 mL Non-sterile 96
14-222-424 240 mL Sterile 96
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
This item is not returnable. View return policy
Catalog No. 14-222-422 Supplier Axygen™ Supplier No. RESSW8HPSI
  • $465.50
    $328.00 / Case of 25

    Save $137.50

    30% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Manufactured with virgin polypropylene for excellent chemical resistance

Corning offers a complete line of Axygen single-well and multiple-well reagent reservoirs which are available in multiple formats for automated and manual use. All reservoirs are designed to comply with standard ANSI/SLAS footprint dimensions for microplates and are compatible with most automation workstations. Reagent reservoirs are manufactured with polypropylene for excellent chemical resistance.

  • Durable and cost effective
  • Various formats for multiple applications
TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance ANSI/SLAS, RNase/DNase-free, non-pyrogenic
Color Clear
Capacity (Metric) 290 mL
Material Polypropylene
Sterility Sterile
Type Reagent Reservoir
No. per Case 25/Cs.
No. per Pack 1/Pk.
Packaging Individually Wrapped; 25/Cs.
Channels 8
Disposable Single-use
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.