Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Axygen™ Single Well Low Profile Reagent Reservoirs
GREENER_CHOICE

Catalog No. 14222420 Shop All Axygen Products
Change view
Click to view available options
Capacity (Metric):
86 mL
90 mL
92 mL
Sterility:
Non-sterile
Sterile
Channels:
1
96
384
Packaging:
25/Cs.
Individually Wrapped; 25/Cs.
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric) Sterility Channels Packaging
14-222-420 92 mL Sterile 384 Individually Wrapped; 25/Cs.
14-222-425 86 mL Non-sterile 96 25/Cs.
14-222-419 92 mL Non-sterile 384 25/Cs.
14-222-426 86 mL Sterile 96 Individually Wrapped; 25/Cs.
14-222-413 90 mL Non-sterile 1 25/Cs.
14-222-414 90 mL Sterile 1 Individually Wrapped; 25/Cs.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 14-222-420 Supplier Axygen™ Supplier No. RESSW384LPSI
  • PRODUCT PROMOTION
    $532.50
    $281.00 / Case of 25

    Save $251.50

    47% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
View all promotions
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Manufactured with virgin polypropylene for excellent chemical resistance

Corning offers a complete line of Axygen single-well and multiple-well reagent reservoirs which are available in multiple formats for automated and manual use. All reservoirs are designed to comply with standard ANSI/SLAS footprint dimensions for microplates and are compatible with most automation workstations. Reagent reservoirs are manufactured with polypropylene for excellent chemical resistance.

  • Durable and cost effective
  • Various formats for multiple applications
TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance ANSI/SLAS, RNase/DNase-free, non-pyrogenic
Color Clear
Capacity (Metric) 92 mL
Material Polypropylene
Sterility Sterile
Type Reagent Reservoir
Packaging Individually Wrapped; 25/Cs.
Channels 384
Disposable Single-use
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.