Learn More
Heathrow Scientific™ Single Wrapped Dual Reagent Reservoir
Description
- Divided reservoir design allows pipetting of two different regents or maximizes recovery of using only single channel and small amount of reagent
- Trough design with a v shaped bottom helps ensure that all the reagent is accessible and can be easily aspirated
- Thick wall construction and wide base adds stability and durability making the reservoir easy to handle without product deformation which results in less spillage
- Reservoirs can be conveniently stacked with a unique nesting design that enables easy separation
- Designed to stay flat on the benchtop and not warp
- Sterile, sealed in bags and electron beam sterilized
- DNase, RNase and Pyrogen free
- Sealed in easy-to-open bags
- Quality certificate available on request
- Disposable
Specifications
Specifications
| Capacity (English) | 0.85 oz. |
| Color | White |
| Capacity (Metric) | 25 mL |
| Dimensions (L x W x H) | 4 x 1 x 0.8 in. |
| Material | Polystyrene |
| Sterility | Sterile |
| Type | Reservoir |
| For Use With (Application) | Liquid Samples |
| Packaging | Single Wrapped |
| Quantity | 1/Bag, 100 Bags/Pk. |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.