Learn More
Crescent Chemical Co Inc SINGLEQUANT ASSAY KIT
SDP
Catalog No. 50248357
Encompass_Preferred
SingleQuant Assay Kit Single tube format assay kit for protein quantification. The assay bases on the precipitation of proteins as insoluble dye complexes with acidic, ethanolic amido black 10B solution (1,2). After precipitation the protein-dye complexes are spinned
Catalog No. 50-248-357 Supplier Crescent Chemical Co Inc Supplier No. SE39226.01
SingleQuant Assay Kit Single tube format assay kit for protein quantification. The assay bases on the precipitation of proteins as insoluble dye complexes with acidic, ethanolic amido black 10B solution (1,2). After precipitation the protein-dye complexes are spinned