Learn More
Lonza Walkersville REGM Renal Epithelial Cell Growth Medium SingleQuots Kit
Catalog No. NC9411806
Encompass
REGM SingleQuots Kit are components sold separately and does not include the basal medium. These are part of the REGM Renal Epithelial Cell Growth Medium BulletKit (Catalog Number CC-3190). The full REGM BulletKit consisting of basal medium and supplements can be purchased together using the catalog number CC-3190.REGM Renal Epithelial Cell Growth Medium BulleKit is formulated to maintain and expand most renal epithelial cells. REGM contains 0.5% serum.Content:1 x REGM SingleQuots Supplement Pack (CC-4127) containing:1 x Green Cap Vial with hEGF, 0.50 mL1 x Lilac Cap Vial with Insulin, 0.50 mL1 x Natural Cap Vial with Hydrocortisone, 0.50 mL1 x Red Cap Vial with GA-1000, 0.50 mL1 x Bottle with FBS, 2.50 mL1 x Natural Cap Vial with Transferrin, 0.50 mL1 x Amber Vial with Triiodothyronine, 0.50 mL1 x Amber Vial with Epinephrine, 0.50 mL
Catalog No. NC9411806 Supplier Lonza Walkersville Supplier No. CC4127
REGM SingleQuots Kit are components sold separately and does not include the basal medium. These are part of the REGM Renal Epithelial Cell Growth Medium BulletKit (Catalog Number CC-3190). The full REGM BulletKit consisting of basal medium and supplements can be purchased together using the catalog number CC-3190.REGM Renal Epithelial Cell Growth Medium BulleKit is formulated to maintain and expand most renal epithelial cells. REGM contains 0.5% serum.Content:1 x REGM SingleQuots Supplement Pack (CC-4127) containing:1 x Green Cap Vial with hEGF, 0.50 mL1 x Lilac Cap Vial with Insulin, 0.50 mL1 x Natural Cap Vial with Hydrocortisone, 0.50 mL1 x Red Cap Vial with GA-1000, 0.50 mL1 x Bottle with FBS, 2.50 mL1 x Natural Cap Vial with Transferrin, 0.50 mL1 x Amber Vial with Triiodothyronine, 0.50 mL1 x Amber Vial with Epinephrine, 0.50 mL