Learn More
Hach Company SINGLET™ Single-Use pH Buffer Kit, pH 4.01 & 7.00, pk/2x10
Catalog No. 501996657 Shop All Hach Company Products
Encompass
pH Buffer Standard Solution for pH electrode calibration. Sealed foil pouches of pH buffers. pH is ±0.02 at 25°C. Pack of 10 of pH 4.01 and 10 of pH 7.00, 20 mL each
Catalog No. 50-199-6657 Supplier Hach Company Supplier No. 2769920
pH Buffer Standard Solution for pH electrode calibration. Sealed foil pouches of pH buffers. pH is ±0.02 at 25°C. Pack of 10 of pH 4.01 and 10 of pH 7.00, 20 mL each