Learn More
Hach Company SINGLET™ Single-Use pH Buffer Kit, pH 7.00 & 10.01, pk/2x10
Catalog No. 501996656 Shop All Hach Company Products
Encompass
pH Buffer Standard Solution for pH electrode calibration. Sealed foil pouches of pH buffers. pH is ±0.02 at 25°C. Pack of 10 of pH 7.00 and 10 of pH 10.01, 20 mL each
Catalog No. 50-199-6656 Supplier Hach Company Supplier No. 2769820
pH Buffer Standard Solution for pH electrode calibration. Sealed foil pouches of pH buffers. pH is ±0.02 at 25°C. Pack of 10 of pH 7.00 and 10 of pH 10.01, 20 mL each