Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SINGLET™ Single-Use pH Buffer, pH 7.00 (NIST), Hach Lange™

Catalog No. 501996659 Shop All Hach Company Products
Encompass
Change view
Click to view available options
Quantity:
20 Pk.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-199-6659 20 Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-199-6659 Supplier Hach Company Supplier No. 2770120
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Hach Lange SINGLET Single-Use pH Buffer, pH 7.00 (NIST), Quantity: 20 pk., Yellow, pH: 7

TRUSTED_SUSTAINABILITY

Specifications

Color Yellow
pH 7
Physical Form Solid
Quantity 20 Pk.
Solution Type Buffer
Formulation Water, Dipotassium Phosphate, Sodium Chloride, Disodium Orthophosphate Heptahydrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.