Learn More
Hach Company Singlet™ Single-use Conductivity Standard Solution, 12.88 mS/cm, KCl, 20 mL
Catalog No. 501996664 Shop All Hach Company Products
Encompass
Conductivity Standard Solution. Sealed foil pouches of 12.88 uS/cm ±10% at 25 °C. Potassium Chloride (KCl). Ready for use, 20 mL each
Catalog No. 50-199-6664 Supplier Hach Company Supplier No. 2771520
Conductivity Standard Solution. Sealed foil pouches of 12.88 uS/cm ±10% at 25 °C. Potassium Chloride (KCl). Ready for use, 20 mL each