Learn More
Hach Company Singlet™ Single-use Conductivity Standard Solution, 147 µS/cm, KCl, 20 mL
Catalog No. 501996662 Shop All Hach Company Products
Encompass
Conductivity Standard Solution. Sealed foil pouches of 147 uS/cm ±10% at 25 °C. Potassium Chloride (KCl). Ready for use, 20 mL each
Catalog No. 50-199-6662 Supplier Hach Company Supplier No. 2771320
Conductivity Standard Solution. Sealed foil pouches of 147 uS/cm ±10% at 25 °C. Potassium Chloride (KCl). Ready for use, 20 mL each