Learn More
Hach Company SINGLET™ Single-Use pH Buffer, pH 4.01, pk/20
Supplier:  Hach Company 2770020
Encompass
pH Buffer Standard Solution for pH electrode calibration. Sealed foil pouches of pH buffers. pH is ±0.02 at 25°C. Pack of 20 of pH 4.01, 20 mL each
Catalog No. 50-199-6658