Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sinigrin Hydrate 98.0+%, TCI America™
SDP

Catalog No. S0156100MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S0156100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S0156100MG Supplier TCI America Supplier No. S0156100MG
Add to Cart
Add to Cart

Chemical Identifiers

CAS 3952-98-5
Molecular Formula C10H16KNO9S2
Molecular Weight (g/mol) 397.454
MDL Number MFCD00006616
InChI Key QKFAFSGJTMHRRY-IMCRRPDGSA-M
Synonym potassium myronate, prop-2-enyl glucosinolate
PubChem CID 122130679
IUPAC Name potassium;[(E)-1-[(2R,3S,4R,5R,6S)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]sulfanylbut-3-enylideneamino] sulfate
SMILES C=CCC(=NOS(=O)(=O)[O-])SC1C(C(C(C(O1)CO)O)O)O.[K+]

Specifications

Melting Point 127°C
Quantity 100 mg
Formula Weight 397.45
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Sinigrin Hydrate
RTECSNumber LZ5778000
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.