Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sinigrin Monohydrate, Synthetic 98%, Thermo Scientific™

Catalog No. AC291442500
Change view
Click to view available options
Quantity:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC291442500 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC291442500 Supplier Thermo Scientific Chemicals Supplier No. 291442500

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3952-98-5
Molecular Formula C10H16KNO9S2
Molecular Weight (g/mol) 397.454
InChI Key QKFAFSGJTMHRRY-IMCRRPDGSA-M
Synonym potassium myronate, prop-2-enyl glucosinolate
PubChem CID 122130679
IUPAC Name potassium;[(E)-1-[(2R,3S,4R,5R,6S)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]sulfanylbut-3-enylideneamino] sulfate
SMILES C=CCC(=NOS(=O)(=O)[O-])SC1C(C(C(C(O1)CO)O)O)O.[K+]

Specifications

Melting Point 127°C to 130°C
Color White
Elemental Analysis 97.5% min.
Infrared Spectrum Authentic
Quantity 250 mg
Merck Index 14, 8545
Formula Weight 415.48
Physical Form Fine Crystalline Powder
Chemical Name or Material Sinigrin monohydrate, 98%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.