Learn More
Cayman Chemical Sinigrin (hydrate), Purity: ≥95%, 500 mg
SDP
Catalog No. 502935898
Encompass_Preferred
A glucosinolate; inhibits NLRP3 inflammasome activation in ATP-stimulated, LPS-primed RAW 264.7 cells in a concentration-dependent manner; reduces LPS-induced NO and PGE2 production, as well as NF-κB activation, in RAW 264.7 cells; induces cell cycle arrest at the G0/G1 phase in HepG2 hepatocellular carcinoma cells at 0.1 and 0.5 mM; increases in renal glomerular basement membrane thickness, as well as increases in systolic and diastolic blood pressure, in a rat model of angiotensin II-induced hypertension
Catalog No. 50-293-5898 Supplier Cayman Chemical Supplier No. 26028500 MG
A glucosinolate; inhibits NLRP3 inflammasome activation in ATP-stimulated, LPS-primed RAW 264.7 cells in a concentration-dependent manner; reduces LPS-induced NO and PGE2 production, as well as NF-κB activation, in RAW 264.7 cells; induces cell cycle arrest at the G0/G1 phase in HepG2 hepatocellular carcinoma cells at 0.1 and 0.5 mM; increases in renal glomerular basement membrane thickness, as well as increases in systolic and diastolic blood pressure, in a rat model of angiotensin II-induced hypertension